2,4,6-Trimethyl-N-(4-(trifluormethyl)benzyl)benzolsulfonamid ≥97%
|
Achtung
|
H55384.03EA
395
EUR
H55384.03
2,4,6-Trimethyl-N-(4-(trifluormethyl)benzyl)benzolsulfonamid ≥97%
2,4,6-Trimethyl-N-(4-(trifluormethyl)benzyl)benzolsulfonamid
Formel:
C₁₇H₁₈F₃NO₂S Molekulargewicht: 357,39 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01652040 CAS-Nummer: 305849-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...