5-Amino-3-(4-chlorphenyl)isoxazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL644943-1GEA 99.3 EUR
SIAL644943-1G
5-Amino-3-(4-chlorphenyl)isoxazol, Sigma-Aldrich®
5-Amino-3-(4-chlorophenyl)isoxazole
Formel: C₉H₇ClN₂O
Molekulargewicht: 194,6177 g/mol
MDL Number: MFCD01569450
CAS-Nummer: 33866-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR