5-(Methylthio)-1H-tetrazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL448257-1GEA 22.7 EUR
SIAL448257-1G
5-(Methylthio)-1H-tetrazol, Sigma-Aldrich®
5-(Methylthio)-1H-tetrazole
Formel: C₂H₄N₄S
MDL Number: MFCD00068726
CAS-Nummer: 29515-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR