5-(Benzylthio)-1H-tetrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: BTTBMT

SIAL75666-5G-FEA 163 EUR
SIAL75666-5G-F
5-(Benzylthio)-1H-tetrazol, Sigma-Aldrich®
5-(Benzylthio)-1H-tetrazol
Formel: C₈H₈N₄S
Molekulargewicht: 192,24 g/mol
MDL Number: MFCD00068731
CAS-Nummer: 21871-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR