3-Carboxy-2-methoxyphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H53323.03EA
390
EUR
H53323.03
3-Carboxy-2-methoxyphenylboronsäure ≥98%
3-Carboxy-2-methoxyphenylboronsäure
Formel:
C₈H₉BO₅ Molekulargewicht: 195,97 g/mol Schmelzpunkt: 152…155 °C |
MDL Number:
MFCD09027237 CAS-Nummer: 913836-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...