2-Chlor-4-(methylcarbamoyl)phenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H55778.03EA
864
EUR
H55778.03
H55778.MD
2-Chlor-4-(methylcarbamoyl)phenylboronsäure ≥97%
2-Chlor-4-(methylcarbamoyl)phenylboronsäure
Formel:
C₈H₉BClNO₃ Molekulargewicht: 213,43 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22125174 CAS-Nummer: 1451391-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...