3-Chlor-4-(tert-butylcarbamoyl)phenylboronsäure ≥98%
|
Achtung
|
H53069.03EA
277
EUR
H53069.03
3-Chlor-4-(tert-butylcarbamoyl)phenylboronsäure ≥98%
3-Chlor-4-(tert-butylcarbamoyl)phenylboronsäure
Formel:
C₁₁H₁₅BClNO₃ Molekulargewicht: 255,51 g/mol Schmelzpunkt: 130…134 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07363771 CAS-Nummer: 850589-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...