2-Amino-5-nitrothiobenzamid 97%
Lieferant: Thermo Scientific
|
Achtung
|
H52237.06EA
181
EUR
H52237.06
H52237.03
2-Amino-5-nitrothiobenzamid 97%
2-Amino-5-nitrothiobenzamid
Formel:
C₇H₇N₃O₂S Molekulargewicht: 197,21 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02258912 CAS-Nummer: 25026-97-5 EINECS: 246-572-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...