Sulfaethoxypyridazin, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

02743-50MGEA 376 EUR
02743-50MG
Sulfaethoxypyridazin, Supelco®
Sulfaethoxypyridazin
Formel: C₁₂H₁₄N₄O₃S
MDL Number: MFCD00869228
CAS-Nummer: 963-14-4
EINECS: 213-514-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR