Sulfamethoxypyridazin
PROOICRS0420EA
0
EUR
PROOICRS0420
Sulfamethoxypyridazin
Sulfamethoxypyridazin
Formel:
C₁₁H₁₂N₄O₃S Molekulargewicht: 280,3 g/mol |
MDL Number:
MFCD00057372 CAS-Nummer: 80-35-3 EINECS: 201-272-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...