2,5-Dimethoxybenzolsulfonamid
Lieferant: Thermo Scientific
MAYBTL00904.10EA
0
EUR
MAYBTL00904.10
2,5-Dimethoxybenzolsulfonamid
2,5-Dimethoxybenzolsulfonamid
Formel:
C₈H₁₁NO₄S Molekulargewicht: 217,25 g/mol |
MDL Number:
MFCD00833415 CAS-Nummer: 19116-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...