4-Brom-2-methylthiobenzamid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL684740-1GEA
61.9
EUR
SIAL684740-1G
4-Brom-2-methylthiobenzamid, Sigma-Aldrich®
4-Brom-2-methylthiobenzamid
Formel:
C₈H₈BrNS Molekulargewicht: 230,12 g/mol |
MDL Number:
MFCD09025669 CAS-Nummer: 397845-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...