1-Benzyloxy-2-methyl-3-nitrobenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL496804-5GEA 106 EUR
SIAL496804-5G
1-Benzyloxy-2-methyl-3-nitrobenzol, Sigma-Aldrich®
1-Benzyloxy-2-methyl-3-nitrobenzol
Formel: C₁₄H₁₃NO₃
Molekulargewicht: 243,26 g/mol
MDL Number: MFCD00100652
CAS-Nummer: 20876-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR