Isochinolin-5-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

SIAL675180-1GEA 113 EUR
SIAL675180-1G
Isochinolin-5-carbaldehyd, Sigma-Aldrich®
Isochinolin-5-carbaldehyd
Formel: C₁₀H₇NO
Molekulargewicht: 157,17
MDL Number: MFCD03412483
CAS-Nummer: 80278-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR