6-Methoxynicotinonitril
Lieferant: Thermo Scientific
MAYBSPB04785DAEA
0
EUR
MAYBSPB04785DA
MAYBSPB04785.10
6-Methoxynicotinonitril
6-Methoxynicotinonitril
Formel:
C₇H₆N₂O Molekulargewicht: 134,1400 g/mol |
MDL Number:
MFCD00084981 CAS-Nummer: 15871-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...