5-(Chlormethyl)-3-(4-chlorphenyl)-1,2,4-oxadiazol
Lieferant: Thermo Scientific
MAYBCD02742DAEA
0
EUR
MAYBCD02742DA
MAYBCD02742CB
5-(Chlormethyl)-3-(4-chlorphenyl)-1,2,4-oxadiazol
5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole
Formel:
C₉H₆Cl₂N₂O |
MDL Number:
MFCD00119077 CAS-Nummer: 57238-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...