3-Amino-2-chlor-5-methylpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL701882-5GEA 86.3 EUR
SIAL701882-5G
3-Amino-2-chlor-5-methylpyridin, Sigma-Aldrich®
3-Amino-2-chloro-5-methylpyridine
Formel: C₆H₇ClN₂
Molekulargewicht: 142,5900 g/mol
MDL Number: MFCD03427656
CAS-Nummer: 34552-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR