5,6,7,8-Tetrahydrochinoxalin

Lieferant: Thermo Scientific

MAYBJFD01263.1EA 0 EUR
MAYBJFD01263.1 MAYBJFD01263.50 MAYBJFD01263.10
5,6,7,8-Tetrahydrochinoxalin
5,6,7,8-Tetrahydrochinoxalin
Formel: C₈H₁₀N₂
Molekulargewicht: 134,18 g/mol
MDL Number: MFCD00038078
CAS-Nummer: 34413-35-9
EINECS: 252-002-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR