5,6,7,8-Tetrahydrochinoxalin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ300
SIALW332100-100G-KEA
412
EUR
SIALW332100-100G-K
SIALW332100-25G-K
SIALW332100-1KG-K
5,6,7,8-Tetrahydrochinoxalin, Sigma-Aldrich®
5,6,7,8-Tetrahydrochinoxalin
Formel:
C₈H₁₀N₂ Molekulargewicht: 134,18 g/mol |
MDL Number:
MFCD00038078 CAS-Nummer: 34413-35-9 EINECS: 252-002-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...