tert-Butyl-4-isothiocyanatobenzoat, Sigma-Aldrich®

SIAL59816-1G-FEA 202 EUR
SIAL59816-1G-F
tert-Butyl-4-isothiocyanatobenzoat, Sigma-Aldrich®
tert-Butyl-4-isothiocyanatobenzoat
Formel: C₁₂H₁₃NO₂S
MDL Number: MFCD03453226
CAS-Nummer: 486415-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR