6-Brom-2-pyridincarbonitril, Sigma-Aldrich®

Lieferant: Merck

SIAL684902-1GEA 73.5 EUR
SIAL684902-1G
6-Brom-2-pyridincarbonitril, Sigma-Aldrich®
6-Brom-2-pyridincarbonitril
Formel: C₆H₃BrN₂
MDL Number: MFCD00234142
CAS-Nummer: 122918-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR