5-Chlorbenzoxazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL666602-5GEA 62 EUR
SIAL666602-5G
5-Chlorbenzoxazol, Sigma-Aldrich®
5-Chlorobenzoxazole
Formel: C₇H₄ClNO
Molekulargewicht: 153,57
MDL Number: MFCD00216939
CAS-Nummer: 17200-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR