Ethylenglycol-[D2] (98% D), Sigma-Aldrich®
|
Achtung
|
M-Clarity™ quality level = MQ200
SIAL343811-1GEA
98
EUR
SIAL343811-1G
Ethylenglycol-[D2] (98% D), Sigma-Aldrich®
Ethylenglycol-[D2]
Formel:
C₂H₄D₂O₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00075277 CAS-Nummer: 2219-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...