4-Propoxybenzonitril
Lieferant: Thermo Scientific
MAYBBTB09860DAEA
0
EUR
MAYBBTB09860DA
MAYBBTB09860EA
MAYBBTB09860.50
4-Propoxybenzonitril
4-Propoxybenzonitril
Formel:
C₁₀H₁₁NO |
MDL Number:
MFCD00173874 CAS-Nummer: 60758-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...