4-(Ethylamino)benzoesäure

Lieferant: Thermo Scientific

MAYBBTB10315DAEA 0 EUR
MAYBBTB10315DA MAYBBTB10315CB
4-(Ethylamino)benzoesäure
4-(Ethylamino)benzoic acid
Formel: C₉H₁₁NO₂
MDL Number: MFCD00102118
CAS-Nummer: 7409-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR