4-Isobutylbenzoesäure
Lieferant: LGC Standards PROMOCHEM
PROOMM0002.03EA
1660
EUR
PROOMM0002.03
4-Isobutylbenzoesäure
4-Isobutylbenzoesäure
Formel:
C₁₁H₁₄O₂ Molekulargewicht: 178,2277 g/mol |
MDL Number:
MFCD00191658 CAS-Nummer: 38861-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...