Methyl-2,6-dichlorisonicotinat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL675962-5GEA
352
EUR
SIAL675962-5G
Methyl-2,6-dichlorisonicotinat, Sigma-Aldrich®
Methyl-2,6-dichlorisonicotinat
Formel:
C₇H₅Cl₂NO₂ Molekulargewicht: 206,03 g/mol |
MDL Number:
MFCD00044409 CAS-Nummer: 42521-09-5 EINECS: 255-868-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...