Aluminiumchlorid Hydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL229393-100GEA
220
EUR
SIAL229393-100G
SIAL229393-25G
Aluminiumchlorid Hydrat, Sigma-Aldrich®
Aluminiumchlorid Hydrat
Formel:
AlCl₃·H₂O Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
10124-27-3 EINECS: 231-208-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...