Methyl-3-amino-2-methylbenzoat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL530271-25GEA 141 EUR
SIAL530271-25G
Methyl-3-amino-2-methylbenzoat, Sigma-Aldrich®
Methyl-3-amino-2-methylbenzoat
Formel: C₉H₁₁NO₂
Molekulargewicht: 165,19 g/mol
MDL Number: MFCD00211328
CAS-Nummer: 18583-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR