3-(4-Methylphenyl)-5-phenyl-1,2,4-oxadiazol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H54813.03EA
117
EUR
H54813.03
H54813.MD
3-(4-Methylphenyl)-5-phenyl-1,2,4-oxadiazol ≥97%
3-(4-Methylphenyl)-5-phenyl-1,2,4-oxadiazole
Formel:
C₁₅H₁₂N₂O Molekulargewicht: 236,27 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02650635 CAS-Nummer: 16151-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...