Filter tips for Liquidator™ 96, BioClean™ LTS™
Lieferant: Mettler-Toledo
RAIN17010646EA 411 EUR
RAIN17010646
Filter tips for Liquidator™ 96, BioClean™ LTS™
Pipettenspitzen
These filter tips are designed for use with the Liquidator™ 96 manual pipetting system.
  • Hydrophobic filter forms a solid barrier against aerosols and liquids
  • No contamination, no additives, no flakes, just pure protection
  • 100% free of DNA, DNAse, RNAse, ATP, Pyrogens, PCR inhibitors, additives and bioactive contaminants
  • Perfect production and advanced LTS design
  • No deformations, no cracks, no flash