Flame-retardant shirts, Nomex® Comfort
Lieferant: Fristads Kansas Group
KWIN10092153049EA 132.9 EUR
KWIN10092153049 KWIN10092153037 KWIN10092153045 KWIN10092153050
Flame-retardant shirts, Nomex® Comfort
Men's shirt with long sleeves, made of 95% Nomex® and 5% Kevlar®.
  • Feels soft like a traditional textile fabric
  • Lightweight and comfortable, ideal for every-day wear
  • Plastic buttons
  • Two chest pockets with flaps
Weight: 180 g/m²
Zertifizierungen: EN 531 (class A, B1 and C1)