Cyclopentadienylmolybdäntricarbonyl dimer, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL117609-1GEA 46.7 EUR
SIAL117609-1G
Cyclopentadienylmolybdäntricarbonyl dimer, Sigma-Aldrich®
Cyclopentadienylmolybdäntricarbonyl dimer
Formel: C₁₆H₁₀Mo₂O₆
Molekulargewicht: 490,13 g/mol
MDL Number: MFCD00009947
CAS-Nummer: 12091-64-4
EINECS: 235-156-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR