2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin 96%

Lieferant: Thermo Scientific
Gefahr

H51574.03EA 476 EUR
H51574.03
2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin 96%
2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin
Formel: C₁₇H₃₂N₂O₂SSn
Molekulargewicht: 447,22 g/mol
MDL Number: MFCD07787379
CAS-Nummer: 122476-85-1
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR