2-Ethoxy-4-(tributylstannyl)thiazol 96%
Lieferant: Thermo Scientific
|
Gefahr
|
H51505.MDEA
93.3
EUR
H51505.MD
2-Ethoxy-4-(tributylstannyl)thiazol 96%
2-Ethoxy-4-(tributylstannyl)thiazol
Formel:
C₁₇H₃₃NOSSn Molekulargewicht: 418,21 g/mol |
MDL Number:
MFCD07787373 CAS-Nummer: 240816-28-8 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...