5-(Tributylstannyl)thiazol

Gefahr

MAYBMO08371CBEA 0 EUR
MAYBMO08371CB
5-(Tributylstannyl)thiazol
5-(Tributylstannyl)thiazol
Formel: C₁₅H₂₉NSSn
MDL Number: MFCD01319074
CAS-Nummer: 157025-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR