4-(2-Chlor-5-methylphenoxy)piperidin ≥99%
|
Achtung
|
H50778.MDEA
71
EUR
H50778.MD
4-(2-Chlor-5-methylphenoxy)piperidin ≥99%
4-(2-Chlor-5-methylphenoxy)piperidin
Formel:
C₁₂H₁₆ClNO Molekulargewicht: 225,72 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06246346 CAS-Nummer: 254883-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...