4-[2-(Trifluormethoxy)phenoxy]piperidin Hydrochlorid 95%
Lieferant: Thermo Scientific
|
Achtung
|
H51178.03EA
386
EUR
H51178.03
4-[2-(Trifluormethoxy)phenoxy]piperidin Hydrochlorid 95%
4-[2-(Trifluormethoxy)phenoxy]piperidin Hydrochlorid
Formel:
C₁₂H₁₅ClF₃NO₂ Molekulargewicht: 297,71 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11870723 CAS-Nummer: 950649-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...