5-Cyanphthalid

Lieferant: Molekula

MOLEM50675324EA 0 EUR
MOLEM50675324
5-Cyanphthalid
5-Cyanphthalid
Formel: C₉H₅NO₂
Molekulargewicht: 159,1415 g/mol
MDL Number: MFCD01072882
CAS-Nummer: 82104-74-3
EINECS: 279-900-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR