5-Cyanphthalid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL647195-5GEA 35 EUR
SIAL647195-5G SIAL647195-25G
5-Cyanphthalid, Sigma-Aldrich®
5-Cyanphthalid
Formel: C₉H₅NO₂
MDL Number: MFCD01072882
CAS-Nummer: 82104-74-3
EINECS: 279-900-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR