4-Chlor-3-(trifluormethyl)pyridin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL680036-250MGEA 61.5 EUR
SIAL680036-250MG
4-Chlor-3-(trifluormethyl)pyridin Hydrochlorid, Sigma-Aldrich®
4-Chlor-3-(trifluormethyl)pyridin Hydrochlorid
Formel: C₆H₄Cl₂F₃N
MDL Number: MFCD04972770
CAS-Nummer: 732306-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR