1-(3,5-Dichlor-4-pyridinyl)piperazin
Lieferant: Thermo Scientific
MAYBSPB06834DAEA
0
EUR
MAYBSPB06834DA
1-(3,5-Dichlor-4-pyridinyl)piperazin
1-(3,5-Dichlor-4-pyridinyl)piperazin
Formel:
C₉H₁₁Cl₂N₃ Molekulargewicht: 232,11 g/mol |
MDL Number:
MFCD00221400 CAS-Nummer: 175277-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...