4-Pyridinacetamid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
B24734.06EA
135
EUR
B24734.06
B24734.03
4-Pyridinacetamid ≥98%
4-Pyridinacetamid
Formel:
C₇H₈N₂O Schmelzpunkt: 144…147 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02685124 CAS-Nummer: 39640-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...