2-(2,4-Dichlorphenoxy)benzonitril

Lieferant: Thermo Scientific

MAYBCD08873.10EA 0 EUR
MAYBCD08873.10
2-(2,4-Dichlorphenoxy)benzonitril
2-(2,4-Dichlorphenoxy)benzonitril
Formel: C₁₃H₇Cl₂NO
MDL Number: MFCD00052459
CAS-Nummer: 175136-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR