Propargylbenzolsulfonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL325341-100MLEA 227 EUR
SIAL325341-100ML
Propargylbenzolsulfonat, Sigma-Aldrich®
Propargylbenzolsulfonat
Formel: C₉H₈O₃S
MDL Number: MFCD00013459
CAS-Nummer: 6165-75-9
EINECS: 228-203-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR