2-(Cyclopropylmethoxy)benzonitril

Lieferant: Thermo Scientific

CHMPRH01471.250EA 0 EUR
CHMPRH01471.250 MAYBRH01471.1
2-(Cyclopropylmethoxy)benzonitril
2-(Cyclopropylmethoxy)benzonitril
Formel: C₁₁H₁₁NO
CAS-Nummer: 83728-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR