4-Cyan-4'-hexyloxybiphenyl, Sigma-Aldrich®

Lieferant: Merck

SIAL338656-1GEA 106 EUR
SIAL338656-1G
4-Cyan-4'-hexyloxybiphenyl, Sigma-Aldrich®
4-Cyan-4'-hexyloxybiphenyl
Formel: C₁₉H₂₁NO
Molekulargewicht: 279,38 g/mol
MDL Number: MFCD00134145
CAS-Nummer: 41424-11-7
EINECS: 255-358-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR