4-Cyan-4'-pentyloxybiphenyl, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL328529-1GEA 108 EUR
SIAL328529-1G
4-Cyan-4'-pentyloxybiphenyl, Sigma-Aldrich®
4-Cyan-4'-pentyloxybiphenyl
Formel: C₁₈H₁₉NO
Molekulargewicht: 281,42 g/mol
MDL Number: MFCD00074879
CAS-Nummer: 52364-71-3
EINECS: 257-875-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR