β-Cyclocitral

Lieferant: Molekula

MOLE18573476-25GEA 178 EUR
MOLE18573476-25G
β-Cyclocitral
β-Cyclocitral
Formel: C₁₀H₁₆O
Molekulargewicht: 152,23 g/mol
MDL Number: MFCD00079078
CAS-Nummer: 432-25-7
EINECS: 207-081-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR