β-Cyclocitral, Sigma-Aldrich®
M-Clarity™ Qualitätsstufe = MQ300
SIALW363928-1KG-KEA
1270
EUR
SIALW363928-1KG-K
SIALW363928-100G-K
β-Cyclocitral, Sigma-Aldrich®
β-Cyclocitral
Formel:
C₁₀H₁₆O Molekulargewicht: 152,23 g/mol |
MDL Number:
MFCD00079078 CAS-Nummer: 432-25-7 EINECS: 207-081-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...